missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzophenone 98.0+%, TCI America™

Catalog No. A11405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11405G 5 g
A114025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11405G Supplier TCI America Supplier No. A11405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1137-41-3
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00007895
InChI Key RBKHNGHPZZZJCI-UHFFFAOYSA-N
Synonym 4-aminobenzophenone, 4-aminophenyl phenyl methanone, 4-benzoylaniline, p-aminobenzophenone, methanone, 4-aminophenyl phenyl, benzophenone, 4-amino, p-benzoylaniline, usaf a-233, 4-aminophenyl-phenylmethanone, 4-aminophenyl-phenyl-methanone
PubChem CID 14346
IUPAC Name 4-benzoylaniline
SMILES NC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 123°C
Quantity 5 g
Formula Weight 197.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminobenzophenone
RTECSNumber DJ0175000
TSCA Yes