missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzophenone, 98%

Catalog No. AAA1375718
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1375718 50 g
AAA1375709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1375718 Supplier Thermo Scientific Chemicals Supplier No. A1375718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1137-41-3 | C13H11NO | 197.24 g/mol

4-Aminobenzophenone is associated with 4-benzoylpyridine and it is used as a frequency doubler for semiconductor lasers. It is used in the preparation of 1-[4-[N,N-bis(trimethylsilyl)amino]phenyl]-1-phenylethylene. It is used as a precursor to prepare 1,2,3-triazoles, pyrimidotriazoles, benzothiazole and pyrimidinone derivatives. It is involved in the formation of coordination complexes with chromium(III).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzophenone is associated with 4-benzoylpyridine and it is used as a frequency doubler for semiconductor lasers. It is used in the preparation of 1-[4-[N,N-bis(trimethylsilyl)amino]phenyl]-1-phenylethylene. It is used as a precursor to prepare 1,2,3-triazoles, pyrimidotriazoles, benzothiazole and pyrimidinone derivatives. It is involved in the formation of coordination complexes with chromium(III).

Solubility
Soluble in cold water, alcohol and ether.

Notes
Air and light sensitive. Incompatible with strong acids.

Chemical Identifiers

CAS 1137-41-3
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00007895
InChI Key RBKHNGHPZZZJCI-UHFFFAOYSA-N
Synonym 4-aminobenzophenone, 4-aminophenyl phenyl methanone, 4-benzoylaniline, p-aminobenzophenone, methanone, 4-aminophenyl phenyl, benzophenone, 4-amino, p-benzoylaniline, usaf a-233, 4-aminophenyl-phenylmethanone, 4-aminophenyl-phenyl-methanone
PubChem CID 14346
IUPAC Name (4-aminophenyl)-phenylmethanone
SMILES NC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 121°C to 124°C
Boiling Point 245°C to 247°C (22 mmHg)
Quantity 50 g
Beilstein 389292
Solubility Information Soluble in cold water,alcohol and ether.
Formula Weight 197.24
Percent Purity 98%
Chemical Name or Material 4-Aminobenzophenone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 214-506-6
RTECSNumber DJ0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only