missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzoic Acid 99.0+%, TCI America™

Catalog No. A026925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A026925G 25 g
A0269100G 100 g
A0269500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A026925G Supplier TCI America Supplier No. A026925G

Chemical Identifiers

CAS 150-13-0
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007894
InChI Key ALYNCZNDIQEVRV-UHFFFAOYSA-N
Synonym p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino
PubChem CID 978
ChEBI CHEBI:30753
IUPAC Name 4-aminobenzoic acid
SMILES NC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 189°C
pH 3.5 (0.5% soln.)
Quantity 25 g
Formula Weight 137.14
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminobenzoic Acid
EINECSNumber (3)-1454
RTECSNumber DG1400000
TSCA Yes