missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzamidine dihydrochlorure, 97%

Catalog No. AC228050250
Change view
Click to view available options
Quantité:
5 g
25 g
Emballage:
Bouteille en verre
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité Emballage
AC228050250 25 g Bouteille en verre
AC228050050 5 g Bouteille en verre
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC228050250 Supplier Thermo Scientific Chemicals Supplier No. 228050250
Obtenez jusqu’à 1,800 $ en produits chimiques
View all promotions

missing translation for 'validMainframeMsg'

CAS : 2498-50-2 | C7H9N3·2HCl | 208,09 g/mol

Ce Thermo Scientific Chemicals produit de marque faisait à l’origine partie du portefeuille de produits d’Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit / article original d’Acros Organics ou la référence SKU n’a pas changé dans le cadre de la transition de la marque vers Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2498-50-2
Formule moléculaire C7H9N3·2HCl
Poids moléculaire (g/mol) 208.09
Numéro MDL MFCD00013001
Clé InChI GHEHNICLPWTXJC-UHFFFAOYSA-N
Synonyme 4-aminobenzamidine dihydrochloride, p-aminobenzamidine dihydrochloride, 4-aminobenzenecarboximidamide dihydrochloride, 4-aminobenzimidamide dihydrochloride, 4-aminobenzamidine 2hcl, 4-amidinoaniline dihydrochloride, benzenecarboximidamide, 4-amino-, dihydrochloride, unii-3c7vnu62ag, benzamidine, p-amino-, dihydrochloride, 3c7vnu62ag
PubChem CID 75626
ChEBI CHEBI:10618
Nom de l’IUPAC 4-aminobenzènecarboximidamide; Dihydrochlorure
SOURIRES C1=CC(=CC=C1C(=N)N)N.Cl.Cl

Specifications

Nom ou substance chimique 4-Aminobenzamidine dihydrochloride
CAS % minimum 96.0
CAS % maximal 100.0
Point de fusion >300 °C
Couleur Du blanc au jaune
Spectre infrarouge Authentique
Fourchette de pourcentages d’essai 96% min. (Argentométrie)
Emballage Bouteille en verre
Formule linéaire H2NC6H4C(=NH)NH2·2HCl
Quantité 25 g
Poids de formule 208.09
Pourcentage de pureté 97%
Forme physique Poudre fine cristalline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-692-2

Utilisation uniquement de la recherche RUO –