missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoantipyrine 97.0+%, TCI America™

Catalog No. A1302500G
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1302500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1302500G Supplier TCI America Supplier No. A1302500G

missing translation for 'validMainframeMsg'

Specifications

CAS 83-07-8
Melting Point 108°C
Molecular Formula C11H13N3O
Quantity 500 g
MDL Number MFCD00003145
Synonym 4-aminoantipyrine, ampyrone, metapirazone, aminoantipyrine, solvapyrin-a, aminoazophene, 4-aminophenazone, 4-aminoantipyrene, aminoantipyrin, solnapyrin-a
InChI Key RLFWWDJHLFCNIJ-UHFFFAOYSA-N
SMILES CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N
IUPAC Name 4-amino-1,5-dimethyl-2-phenylpyrazol-3-one
Molecular Weight (g/mol) 203.245
PubChem CID 2151
ChEBI CHEBI:59026
Formula Weight 203.25
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminoantipyrine
Show More Show Less
EINECSNumber (9)-0062
RTECSNumber CD2480000
TSCA Yes