missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoantipyrine, BAKER™, J.T. Baker™

Catalog No. 02002104
Change view
Click to view available options
Quantity:
100 g
Packaging:
Wide Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002104 100 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02002104 Supplier Avantor Supplier No. A63005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A grade of chemicals for laboratory use

Specifications

CAS 83-07-8
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula C11H13N3O
Quantity 100 g
Synonym 4-aminoantipyrine, ampyrone, metapirazone, aminoantipyrine, solvapyrin-a, aminoazophene, 4-aminophenazone, 4-aminoantipyrene, aminoantipyrin, solnapyrin-a
InChI Key RLFWWDJHLFCNIJ-UHFFFAOYSA-N
SMILES CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N
IUPAC Name 4-amino-1,5-dimethyl-2-phenylpyrazol-3-one
Molecular Weight (g/mol) 203.245
PubChem CID 2151
ChEBI CHEBI:59026
Formula Weight 203.24
Chemical Name or Material 4-Aminoantipyrine
Show More Show Less