missing translation for 'onlineSavingsMsg'
Learn More

4'-Aminoacetophenone, 99%

Catalog No. AAA1321330
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
1000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1321330 250 g
AAA1321314 25 g
AAA1321318 50 g
AAA132130B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1321330 Supplier Thermo Scientific Chemicals Supplier No. A1321330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-92-3 | C8H9NO | 135.17 g/mol

4'-Aminoacetophenone, is used in pharmaceuticals, medicines and other organic products. It is also used as a reagent for the photometric determination of Ce. As pharmaceutical intemediate or in the determination of palladium and vitaminB1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Aminoacetophenone, is used in pharmaceuticals, medicines and other organic products. It is also used as a reagent for the photometric determination of Ce. As pharmaceutical intemediate or in the determination of palladium and vitaminB1.

Solubility
It is soluble in hot water, ethanol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. acids, Incompatible to Acid chlorides, Acid anhydrides, Chloroformates, Strong oxidizing agents, Strong reducing agents. Stable under ordinary conditions.

Chemical Identifiers

CAS 99-92-3
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00007896
InChI Key GPRYKVSEZCQIHD-UHFFFAOYSA-N
Synonym 4'-aminoacetophenone, 4-aminoacetophenone, 1-4-aminophenyl ethanone, p-aminoacetophenone, 4-acetylaniline, p-acetylaniline, ethanone, 1-4-aminophenyl, p-aminoacetylbenzene, 1-4-aminophenyl ethan-1-one, acetophenone, p-amino
PubChem CID 7468
IUPAC Name 1-(4-aminophenyl)ethanone
SMILES CC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 103°C to 108°C
Boiling Point 292°C to 294°C
Quantity 250 g
Beilstein 471493
Merck Index 14,413
Solubility Information It is soluble in hot water,ethanol and ether.
Formula Weight 135.17
Percent Purity 99%
Chemical Name or Material 4'-Aminoacetophenone
Hazard Category H302-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-801-2
RTECSNumber AM5500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only