missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-N-Fmoc-L-phenylalanine, 98%

Catalog No. AAH6250506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6250506 5 g
AAH6250503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6250506 Supplier Thermo Scientific Chemicals Supplier No. H6250506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 95753-56-3 | C24H22N2O4 | 402.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95753-56-3
Molecular Formula C24H22N2O4
Molecular Weight (g/mol) 402.45
MDL Number MFCD00237664
InChI Key VALNSJHHRPSUDO-QFIPXVFZSA-N
Synonym fmoc-4-amino-l-phenylalanine, fmoc-phe 4-nh2-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-aminophenyl propanoic acid, n, a-fmoc-4-amino-l-phenylalanine, fmoc-4-amino-phenylalanine, l-phenylalanine, 4-amino-n-9h-fluoren-9-ylmethoxy carbonyl, 2s-3-4-aminophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-4-ao-l-phenylalanine, fmoc-phe p-nh2-oh, pubchem12464
PubChem CID 7019927
IUPAC Name (2S)-3-(4-aminophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)N)C(=O)O

Specifications

Quantity 5 g
Formula Weight 402.44
Percent Purity 98%
Chemical Name or Material 4-Amino-N-Fmoc-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only