missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-chloro-2-methylmercaptopyrimidine, 97%

Catalog No. AC103390250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103390250 25 g Glass bottle
AC103390050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC103390250 Supplier Thermo Scientific Chemicals Supplier No. 103390250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1005-38-5 | C5H6ClN3S | 175.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1005-38-5
Molecular Formula C5H6ClN3S
Molecular Weight (g/mol) 175.64
MDL Number MFCD00006088
InChI Key ISUXMAHVLFRZQU-UHFFFAOYSA-N
Synonym 4-amino-6-chloro-2-methylthio pyrimidine, 4-amino-6-chloro-2-methylmercaptopyrimidine, 4-pyrimidinamine, 6-chloro-2-methylthio, 6-chloro-2-methylsulfanyl pyrimidin-4-amine, 6-chloro-2-methylthio pyrimidin-4-amine, pyrimidine, 4-amino-6-chloro-2-methylthio, 4-amino-6-chloro-2-methylthiopyrimidine, 6-chloro-2-methylthiopyrimidin-4-ylamine, 6-amino-4-chloro-2-methylmercaptopyrimidine, 6-chloro-2-methylsulfanyl-pyrimidin-4-amine
PubChem CID 70496
IUPAC Name 6-chloro-2-methylsulfanylpyrimidin-4-amine
SMILES CSC1=NC(=CC(=N1)Cl)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 130°C to 134°C
Color Beige to White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 25, 11
Formula Weight 175.64
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-6-chloro-2-methylmercaptopyrimidine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-734-3

RUO – Research Use Only