missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-chloro-1,3-benzenedisulfonamide 98.0+%, TCI America™

Catalog No. A136225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A136225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A136225G Supplier TCI America Supplier No. A136225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121-30-2
Molecular Formula C6H8ClN3O4S2
Molecular Weight (g/mol) 285.717
MDL Number MFCD00007933
InChI Key IHJCXVZDYSXXFT-UHFFFAOYSA-N
Synonym 4-amino-6-chloro-1,3-benzenedisulfonamide, chloraminophenamide, salmid, idorese, chloroaminophenamide, 1,3-benzenedisulfonamide, 4-amino-6-chloro, 5-chloro-2,4-disulfamylaniline, 4-amino-6-chloro-m-benzenedisulfonamide, 3-chloro-4,6-disulfamoylaniline, 5-chloro-2,4-disulfamoylaniline
PubChem CID 67136
ChEBI CHEBI:3602
IUPAC Name 4-amino-6-chlorobenzene-1,3-disulfonamide
SMILES C1=C(C(=CC(=C1Cl)S(=O)(=O)N)S(=O)(=O)N)N

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 285.72
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-6-chloro-1,3-benzenedisulfonamide