missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-4'-nitrodiphenyl Sulfide 98.0+%, TCI America™

Catalog No. A207725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A207725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A207725G Supplier TCI America Supplier No. A207725G
Only null left

Chemical Identifiers

CAS 101-59-7
Molecular Formula C12H10N2O2S
Molecular Weight (g/mol) 246.28
MDL Number MFCD00007881
InChI Key ZBPKGHOGUVVDLF-UHFFFAOYSA-N
Synonym 4-(4-Nitrophenylthio)aniline, 4-Aminophenyl 4-Nitrophenyl Sulfide
PubChem CID 7566
IUPAC Name 4-[(4-nitrophenyl)sulfanyl]aniline
SMILES NC1=CC=C(SC2=CC=C(C=C2)[N+]([O-])=O)C=C1

Specifications

Melting Point 143°C
Quantity 25 g
Formula Weight 246.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-4′-nitrodiphenyl Sulfide
RTECSNumber WP9640000
TSCA No