missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-4'-chlorodiphenyl Ether 98.0+%, TCI America™

Catalog No. C106925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C106925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C106925G Supplier TCI America Supplier No. C106925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101-79-1
Molecular Formula C12H10ClNO
Molecular Weight (g/mol) 219.67
MDL Number MFCD00043925
InChI Key YTISFYMPVILQRL-UHFFFAOYSA-N
Synonym 4-4-chlorophenoxy aniline, 4-amino-4'-chlorodiphenyl ether, benzenamine, 4-4-chlorophenoxy, p-p-chlorophenoxy aniline, 4-4-chlorophenoxy benzenamine, 4-chloro-4'-aminodiphenyl ether, aniline, p-chlorophenoxy, 4-amino-4-chlorodiphenyl ether, 4'-chloro-4-aminobiphenyl ether, aniline, p-p-chlorophenoxy
PubChem CID 7578
IUPAC Name 4-(4-chlorophenoxy)aniline
SMILES NC1=CC=C(OC2=CC=C(Cl)C=C2)C=C1

Specifications

Melting Point 101°C
Color Gray-White
Boiling Point 205°C
Quantity 25 g
Formula Weight 219.67
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-4′-chlorodiphenyl Ether
RTECSNumber BX1770000
TSCA Yes