missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-(trifluoromethyl)benzonitrile 98.0+%, TCI America™

Catalog No. A26441G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26441G 1 g
A26445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26441G Supplier TCI America Supplier No. A26441G

Chemical Identifiers

CAS 327-74-2
Molecular Formula C8H5F3N2
Molecular Weight (g/mol) 186.137
MDL Number MFCD00275473
InChI Key MWLZJOBGDXBMBP-UHFFFAOYSA-N
Synonym 4-amino-3-trifluoromethyl benzonitrile, 2-amino-5-cyanobenzotrifluoride, 2-trifluoromethyl-4-cyanoaniline, 4-cyano-2-trifluoromethyl aniline, 2-amino-5cyano-trifluorotoluene, 3-trifluoromethyl-4-aminobenzonitrile, benzonitrile, 4-amino-3-trifluoromethyl, 4-amino-3-trifluoromethyl-benzonitrile, 4-amino-3-trifluoromethyl benzenecarbonitrile, 4-amino-3-trifluoromethylbenzonitrile;2-amino-5-cyanobenzotrifluoride
PubChem CID 2779488
IUPAC Name 4-amino-3-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1C#N)C(F)(F)F)N

Specifications

Melting Point 62°C
Boiling Point 100°C
Quantity 1 g
UN Number 3439
Formula Weight 186.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-(trifluoromethyl)benzonitrile