missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitropyridine, 96%

Catalog No. AAL1972514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1972514 25 g
AAL1972506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1972514 Supplier Thermo Scientific Chemicals Supplier No. L1972514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1681-37-4 | C5H5N3O2 | 139.114 g/mol

4-Amino-3-nitropyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-nitropyridine is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 1681-37-4
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.114
MDL Number MFCD00160304
InChI Key IUPPEELMBOPLDJ-UHFFFAOYSA-N
Synonym 4-amino-3-nitropyridine, 4-pyridinamine, 3-nitro, 3-nitro-4-pyridinamine, 3-nitro-4-aminopyridine, 3-nitro-pyridin-4-ylamine, 3-nitro-4-pyridylamine, pubchem1277, acmc-209dxo, 3-nitro-4-amino pyridine, 3-nitro-4-pyridinamine #
PubChem CID 548803
IUPAC Name 3-nitropyridin-4-amine
SMILES C1=CN=CC(=C1N)[N+](=O)[O-]

Specifications

Melting Point 201°C to 205°C
Quantity 25 g
Beilstein 383710
Solubility Information Insoluble in water.
Formula Weight 139.11
Percent Purity 96%
Chemical Name or Material 4-Amino-3-nitropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only