missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitrobenzonitrile, 98%

Catalog No. AAB2346314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2346314 25 g
AAB2346303 1 g
AAB2346306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2346314 Supplier Thermo Scientific Chemicals Supplier No. B2346314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6393-40-4 | C7H5N3O2 | 163.14 g/mol

4-Amino-3-nitrobenzonitrile is used in gamma irradiations effect on release of this compound from polyanhydride matrixes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-nitrobenzonitrile is used in gamma irradiations effect on release of this compound from polyanhydride matrixes.

Solubility
Soluble in ethanol and benzene.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 6393-40-4
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD00013373
InChI Key JAHADAZIDZMHOP-UHFFFAOYSA-N
Synonym 3-nitro-4-aminobenzonitrile, 4-cyano-2-nitroaniline, benzonitrile, 4-amino-3-nitro, 4-amino-3-nitro-benzonitrile, 2-nitro-4-cyanoaniline, 4-amino-3-nitrobenzenecarbonitrile, zlchem 228, pubchem4669, 4-ammo-3-nitrobenzonitrile
PubChem CID 595901
IUPAC Name 4-amino-3-nitrobenzonitrile
SMILES NC1=CC=C(C=C1[N+]([O-])=O)C#N

Specifications

Melting Point 159°C to 161°C
Quantity 25 g
UN Number UN3439
Solubility Information Soluble in ethanol and benzene.
Formula Weight 163.14
Percent Purity 98%
Chemical Name or Material 4-Amino-3-nitrobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only