missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. A13535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13535G 5 g
A135325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13535G Supplier TCI America Supplier No. A13535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2486-70-6
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
InChI Key NHFKECPTBZZFBC-UHFFFAOYSA-N
Synonym 3-methyl-4-aminobenzoic acid, 4-amino-m-toluic acid, benzoic acid, 4-amino-3-methyl, 4-amino-3-methyl benzoic acid, 4-amino-3-methyl benzoate, 4-amino-3-methylbenzoicacid, unii-201cxt4h5v, 4-amino-3-methyl-benzoic acid, benzoic acid, 3-methyl-4-amino, 4-amino-3-methylbenzenecarboxylic acid
PubChem CID 75598
IUPAC Name 4-amino-3-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)C(=O)O)N

Specifications

Melting Point 172°C
Quantity 5 g
Formula Weight 151.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-methylbenzoic Acid