missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-methoxybenzoic acid, 98%

Catalog No. AC391460050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC391460050 5 g Glass bottle
AC391460010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC391460050 Supplier Thermo Scientific Chemicals Supplier No. 391460050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2486-69-3 | C8H9NO3 | 167.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2486-69-3
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD00016539
InChI Key JNFGLYJROFAOQP-UHFFFAOYSA-N
Synonym 4-amino-3-methoxy-benzoic acid, 3-methoxy-4-aminobenzoic acid, 4-amino-m-anisic acid, benzoic acid, 4-amino-3-methoxy, o-anisidine-4-carboxylic acid, 4-amino-3-methoxy benzoic acid, timtec-bb sbb008622, 4-amino-3-methoxybenzoic, 4-amino-3-methoxybenzoic acid, 4-amino-3-methoxybenzoicacid
PubChem CID 288057
IUPAC Name 4-amino-3-methoxybenzoic acid
SMILES COC1=C(C=CC(=C1)C(=O)O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 186°C to 188°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3OC6H3(NH2)COOH
Quantity 5 g
Formula Weight 167.16
Percent Purity 98%
Physical Form Powder, Crystals and/or Chunks or Fused Solid
Chemical Name or Material 4-Amino-3-methoxybenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only