missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-iodopyridine 98.0+%, TCI America™

Numéro de catalogue. p-7118001
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A26411G 1 g
A26415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. A26411G Fournisseur TCI America Code fournisseur A26411G
Il en reste null

Identifiants chimiques

CAS 88511-27-7
Molecular Formula C5H5IN2
Molecular Weight (g/mol) 220.013
MDL Number MFCD02663853
InChI Key ZGOCEDVVZKFHSY-UHFFFAOYSA-N
Synonym 4-amino-3-iodopyridine, 3-iodo-4-aminopyridine, 3-iodopyridin-4-ylamine, 3-iodo-pyridin-4-ylamine, 3-iodo-4-pyridylamine, 4-pyridinamine, 3-iodo, pubchem6632, 3-iodo-4-pyridineamine, 4-amino-3-iodo-pyridin, acmc-20a09a
PubChem CID 1516510
IUPAC Name 3-iodopyridin-4-amine
SMILES C1=CN=CC(=C1N)I

Spécifications

Melting Point 98°C
Color White-Yellow
Quantity 1 g
Formula Weight 220.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-iodopyridine