missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydroxybutyric Acid 98.0+%, TCI America™

Catalog No. A03185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03185G 5 g
A031825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A03185G Supplier TCI America Supplier No. A03185G
Only null left

Chemical Identifiers

CAS 924-49-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00008141
InChI Key YQGDEPYYFWUPGO-UHFFFAOYSA-N
Synonym 4-amino-3-hydroxybutyric acid, gabob, 3-hydroxy-gaba, dl-4-amino-3-hydroxybutyric acid, gabomade, gamibetal, gaboril, buksamin, gabimex, gaminal
PubChem CID 2149
ChEBI CHEBI:16080
IUPAC Name 4-amino-3-hydroxybutanoic acid
SMILES C(C(CN)O)C(=O)O

Specifications

Melting Point 218°C
Color White
Quantity 5 g
Formula Weight 119.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-hydroxybutyric Acid
EINECSNumber (9)-0093
RTECSNumber ES7015000
TSCA Yes