missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-fluorobenzoic Acid 98.0+%, TCI America™

Catalog No. A21655G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21655G 5 g
A21651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21655G Supplier TCI America Supplier No. A21655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 455-87-8
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD01660374
InChI Key JSKXHTHMCCDEGD-UHFFFAOYSA-N
Synonym 3-fluoro-4-aminobenzoic acid, benzoic acid, 4-amino-3-fluoro, 4-amino-3-fluorobenzenecarboxylic acid, 4-amino-3-fluoro-benzoic acid, 4-amino-3-fluorobenzoicacid, pubchem3524, wln: zr bf dvq, acmc-209k3i, 3-14-00-01153 beilstein handbook reference, buttpark 44\09-60
PubChem CID 9971
IUPAC Name 4-amino-3-fluorobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)F)N

Specifications

Quantity 5 g
Formula Weight 155.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-fluorobenzoic Acid
RTECSNumber DG2525000
TSCA No