missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-chloropyridine, 97%

Catalog No. AAH3109306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3109306 5 g
AAH3109303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3109306 Supplier Thermo Scientific Chemicals Supplier No. H3109306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19798-77-7 | C5H5ClN2 | 128.559 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19798-77-7
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD03984473
InChI Key KJBKPRMEMJKXDV-UHFFFAOYSA-N
Synonym 4-amino-3-chloropyridine, 3-chloro-pyridin-4-ylamine, 3-chloro-4-pyridinamine, 3-chloro-4-aminopyridine, 3-chloro-4-pyridinylamine, 4-pyridinamine, 3-chloro, 3-chloro-4-pyridinamin, 3-chloro-4-pyridylamine, 4-amino-3-chloro-pyridine, pubchem6653
PubChem CID 581853
IUPAC Name 3-chloropyridin-4-amine
SMILES C1=CN=CC(=C1N)Cl

Specifications

Melting Point 49°C to 53°C
Odor Unpleasant
Quantity 5 g
Formula Weight 128.56
Percent Purity 97%
Chemical Name or Material 4-Amino-3-chloropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only