missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3,5-dichloropyridine 98.0+%, TCI America™

Catalog No. A15455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15455G 5 g
A154525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15455G Supplier TCI America Supplier No. A15455G

Chemical Identifiers

CAS 22889-78-7
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.00
MDL Number MFCD00125023
InChI Key ISIQAMHROGZHOV-UHFFFAOYSA-N
Synonym 4-amino-3,5-dichloropyridine, 3,5-dichloro-4-aminopyridine, 3,5-dichloro-4-pyridinamine, 4-pyridinamine, 3,5-dichloro, 3,5-dichloro-4-amino pyridine, 3,5-dichloro-4-pyridine amine, 3,5-dichloro-pyridin-4-ylamine, 3,5-dichloro-4-pyridylamine, 3,5-dichloropyridin-4-ylamine, 4-amino-3,5-dichloro pyridine
PubChem CID 89888
IUPAC Name 3,5-dichloropyridin-4-amine
SMILES NC1=C(Cl)C=NC=C1Cl

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 5 g
Formula Weight 163.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3,5-dichloropyridine