missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-methoxybenzoic acid, 97%

Catalog No. AAH2741914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2741914 25 g
AAH2741906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2741914 Supplier Thermo Scientific Chemicals Supplier No. H2741914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2486-80-8 | C8H9NO3 | 167.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2486-80-8
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00114479
InChI Key OLJXRTRRJSMURJ-UHFFFAOYSA-N
Synonym 4-amino-o-anisic acid, 4-amino-2-methoxy-benzoic acid, 4-amino-2-methoxybenzenecarboxylic acid, 2-methoxy-4-amino-benzoic acid, 4-amino-2-methoxybenzoicacid, 4-azanyl-2-methoxy-benzoic acid, benzoic acid, 4-amino-2-methoxy, 2-methoxy-4-aminobenzoic acid, acmc-20aicb, pubchem4934
PubChem CID 75599
IUPAC Name 4-amino-2-methoxybenzoic acid
SMILES COC1=C(C=CC(=C1)N)C(=O)O

Specifications

Melting Point ∽150°C (decomposition)
Quantity 25 g
Formula Weight 167.16
Percent Purity 97%
Chemical Name or Material 4-Amino-2-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-632-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only