missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2-dimethylamino-6-hydroxy-5-nitrosopyrimidine 98.0+%, TCI America™
Specifications
Specifications
| Color | Purple-Red |
| Quantity | 1 g |
| Formula Weight | 183.17 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Amino-2-dimethylamino-6-hydroxy-5-nitrosopyrimidine [for Determination of Co(III), Fe(II)] |
Safety and Handling
| TSCA | No |