missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chlorotoluene-5-sulfonic Acid 98.0+%, TCI America™

Catalog No. A028625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A028625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A028625G Supplier TCI America Supplier No. A028625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88-51-7
Molecular Formula C7H8ClNO3S
Molecular Weight (g/mol) 221.655
MDL Number MFCD00035771
InChI Key VRLPHBSFRWMMPW-UHFFFAOYSA-N
Synonym 4-amino-2-chlorotoluene-5-sulfonic acid, 2b acid, red 2b acid, brilliant toning red amine, benzenesulfonic acid, 2-amino-4-chloro-5-methyl, permanent red 2b amine, 6-amino-4-chloro-m-toluenesulfonic acid, unii-9qn6jt928n, ccris 3406, 2-chloro-4-aminotoluene-5-sulfonic acid
PubChem CID 6935
IUPAC Name 2-amino-4-chloro-5-methylbenzenesulfonic acid
SMILES CC1=CC(=C(C=C1Cl)N)S(=O)(=O)O

Specifications

Quantity 25 g
Formula Weight 221.66
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-chlorotoluene-5-sulfonic Acid
EINECSNumber (3)-2024
RTECSNumber XT6330000
TSCA Yes