missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chloro-5-iodopyridine, 95%, Thermo Scientific Chemicals

Catalog No. AAH3392603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3392603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3392603 Supplier Thermo Scientific Chemicals Supplier No. H3392603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 800402-12-4 | Molecular Formula: C5H4ClIN2 | Molecular Weight: 254.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 800402-12-4
Molecular Formula C5H4ClIN2
Molecular Weight (g/mol) 254.46
MDL Number MFCD09834661
InChI Key DEJUUKULVAIMNF-UHFFFAOYSA-N
Synonym 2-chloro-5-iodo-4-pyridinamine, 4-amino-2-chloro-5-iodopyridine, 4-pyridinamine, 2-chloro-5-iodo, 2-chloro-5-iodo-4-aminopyridine, 2-chloro-5-iodo-pyridin-4-ylamine, 2-chloro-5-iodo-pyridine-4-ylamine, 2-chloro-5-iodo-4-pyridylamine, 2-chloro-4-amino-5-iodopyridine, 2-chloro-5-iodo-4-pyridineamine, 2-chloro-5-iodo-pyridin-4-amine
PubChem CID 45480366
IUPAC Name 2-chloro-5-iodopyridin-4-amine
SMILES NC1=CC(Cl)=NC=C1I

Specifications

Melting Point 126°C to 127°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 254.46
Percent Purity 95%
Chemical Name or Material 4-Amino-2-chloro-5-iodopyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only