missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2,6-diphenylphenol 98.0+%, TCI America™
$82.79 - $163.64
Chemical Identifiers
| CAS | 50432-01-4 |
|---|---|
| Molecular Formula | C18H15NO |
| Molecular Weight (g/mol) | 261.324 |
| MDL Number | MFCD00034070 |
| InChI Key | YCOUFOVMXBWYIX-UHFFFAOYSA-N |
| Synonym | 5′C-Amino-2′C-hydroxy-m-terphenyl |
| PubChem CID | 621555 |
| IUPAC Name | 4-amino-2,6-diphenylphenol |
| SMILES | C1=CC=C(C=C1)C2=CC(=CC(=C2O)C3=CC=CC=C3)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A1269500MG
|
TCI America
A1269500MG |
500 mg |
Each for $82.79
|
|
|||||
|
A12691G
|
TCI America
A12691G |
1 g |
Each for $163.64
|
|
|||||
Chemical Identifiers
| 50432-01-4 | |
| 261.324 | |
| YCOUFOVMXBWYIX-UHFFFAOYSA-N | |
| 621555 | |
| C1=CC=C(C=C1)C2=CC(=CC(=C2O)C3=CC=CC=C3)N |
| C18H15NO | |
| MFCD00034070 | |
| 5′C-Amino-2′C-hydroxy-m-terphenyl | |
| 4-amino-2,6-diphenylphenol |
Specifications
| 50432-01-4 | |
| Red-White | |
| MFCD00034070 | |
| 5′C-Amino-2′C-hydroxy-m-terphenyl | |
| C1=CC=C(C=C1)C2=CC(=CC(=C2O)C3=CC=CC=C3)N | |
| 261.324 | |
| 261.32 | |
| Crystalline Powder |
| 148°C | |
| C18H15NO | |
| 500 mg | |
| YCOUFOVMXBWYIX-UHFFFAOYSA-N | |
| 4-amino-2,6-diphenylphenol | |
| 621555 | |
| ≥98.0% (T) | |
| 4-Amino-2,6-diphenylphenol |
Safety and Handling
TSCA : No