accessibility menu, dialog, popup

UserName

CAS RN 50432-01-4

IUPAC Name: 5-amino-3-phenyl-[1,1'-biphenyl]-2-ol
Synonyms: 5-amino-3-phenyl-[1,1'-biphenyl]-2-ol
Molecular Weight (g/mol): 261.32
Molecular Formula: C18H15NO
InChi Key: YCOUFOVMXBWYIX-UHFFFAOYSA-N
SMILES: NC1=CC(C2=CC=CC=C2)=C(O)C(=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results