missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2,6-diphenylphenol 98.0+%, TCI America™
Supplier: TCI America A12691G
Specifications
| 4-Amino-2,6-diphenylphenol | |
| 148°C | |
| C18H15NO | |
| 1 g | |
| YCOUFOVMXBWYIX-UHFFFAOYSA-N | |
| 4-amino-2,6-diphenylphenol | |
| 621555 | |
| ≥98.0% (T) |
| 50432-01-4 | |
| Red-White | |
| MFCD00034070 | |
| 5′C-Amino-2′C-hydroxy-m-terphenyl | |
| C1=CC=C(C=C1)C2=CC(=CC(=C2O)C3=CC=CC=C3)N | |
| 261.324 | |
| 261.32 | |
| Crystalline Powder |
Chemical Identifiers
| 50432-01-4 | |
| 261.324 | |
| YCOUFOVMXBWYIX-UHFFFAOYSA-N | |
| 621555 | |
| C1=CC=C(C=C1)C2=CC(=CC(=C2O)C3=CC=CC=C3)N |
| C18H15NO | |
| MFCD00034070 | |
| 5′C-Amino-2′C-hydroxy-m-terphenyl | |
| 4-amino-2,6-diphenylphenol |
Safety and Handling
TSCA : No