missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,6-dimethylpyridine 98.0+%, TCI America™

Catalog No. A27495G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27495G 5 g
A27491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27495G Supplier TCI America Supplier No. A27495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3512-80-9 | C7H10N2 | 122.171 g/mol

Chemical Identifiers

CAS 3512-80-9
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00130078
InChI Key ZJXMKPARTVOUAM-UHFFFAOYSA-N
PubChem CID 77047
IUPAC Name 2,6-dimethylpyridin-4-amine
SMILES CC1=CC(=CC(=N1)C)N

Specifications

Melting Point 193°C
Color White-Yellow
Boiling Point 246°C
Quantity 5 g
Synonym 4-amino-2,6-dimethylpyridine, 2,6-dimethyl-pyridin-4-ylamine, 2,6-dimethylpyridin-4-ylamine, 4-amino-2,6-lutidine, 4-pyridinamine, 2,6-dimethyl, 2,6-dimethylpyridin-4yl-amine, 2,6-dimethyl-4-pyridinamine, 2,6-dimethyl-4-aminopyridine, pubchem19474, 4-amino-2, 6-lutidine
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2,6-dimethylpyridine
EINECSNumber (9)-0670
TSCA No