missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,6-dimethylpyridine, 98%

Catalog No. AAH6197006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6197006 5 g
AAH6197003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6197006 Supplier Thermo Scientific Chemicals Supplier No. H6197006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3512-80-9 | C7H10N2 | 122.171 g/mol

4-Amino-2,6-dimethylpyridine, is an important raw material and intermediate used in pharmaceuticals, agrochemicals, dyestuff, field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-2,6-dimethylpyridine, is an important raw material and intermediate used in pharmaceuticals, agrochemicals, dyestuff, field.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 3512-80-9
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00130078
InChI Key ZJXMKPARTVOUAM-UHFFFAOYSA-N
PubChem CID 77047
IUPAC Name 2,6-dimethylpyridin-4-amine
SMILES CC1=CC(=CC(=N1)C)N

Specifications

Quantity 5 g
UN Number UN2811
Synonym 4-amino-2,6-dimethylpyridine, 2,6-dimethyl-pyridin-4-ylamine, 2,6-dimethylpyridin-4-ylamine, 4-amino-2,6-lutidine, 4-pyridinamine, 2,6-dimethyl, 2,6-dimethylpyridin-4yl-amine, 2,6-dimethyl-4-pyridinamine, 2,6-dimethyl-4-aminopyridine, pubchem19474, 4-amino-2, 6-lutidine
Solubility Information Slightly soluble in water.
Formula Weight 122.17
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Amino-2,6-dimethylpyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 225-515-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only