missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,6-dihydroxy-5-nitrosopyrimidine 93.0+%, TCI America™

Catalog No. A12025G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A12025G Supplier TCI America Supplier No. A12025G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5442-24-0
Molecular Formula C4H4N4O3
Molecular Weight (g/mol) 156.101
MDL Number MFCD00012340
InChI Key DKPCSXFEWFSECE-UHFFFAOYSA-N
Synonym 4-Amino-5-nitro-2,6-pyrimidinediol, 6-Amino-5-nitrosouracil
PubChem CID 79510
IUPAC Name 6-amino-5-nitroso-1H-pyrimidine-2,4-dione
SMILES C1(=C(NC(=O)NC1=O)N)N=O

Specifications

Color Purple-Red
Quantity 5 g
UN Number 1325
Formula Weight 156.10
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2,6-dihydroxy-5-nitrosopyrimidine