missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,6-dichloropyridine 97.0+%, TCI America™

Catalog No. A23695G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23695G 5 g
A23691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23695G Supplier TCI America Supplier No. A23695G

Chemical Identifiers

CAS 2587-02-2
Molecular Formula C5H5Cl2N2
Molecular Weight (g/mol) 164.01
MDL Number MFCD00052832
InChI Key WAEZOSSWRXDWAX-UHFFFAOYSA-O
Synonym 4-amino-2,6-dichloropyridine, 2,6-dichloro-4-aminopyridine, 2,6-dichloro-pyridin-4-ylamine, 4-pyridinamine, 2,6-dichloro, 2,6-dichloro-4-pyridylamine, 2,6-dichloropyridin-4-ylamine, 2,6-dichloro-4-amino pyridine, 2,6-dichloro-4-pyridinamine, pubchem5530, acmc-1cot2
PubChem CID 137652
IUPAC Name 2,6-dichloropyridin-4-amine
SMILES C1=C(C=C(N=C1Cl)Cl)N

Specifications

Melting Point 175°C
Color Yellow
Quantity 5 g
Formula Weight 163.00
Percent Purity ≥97.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2,6-dichloropyridine