missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,5-difluorobenzonitrile 98.0+%, TCI America™

Catalog No. A21065G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21065G 5 g
A21061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21065G Supplier TCI America Supplier No. A21065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112279-61-5
Molecular Formula C7H4F2N2
Molecular Weight (g/mol) 154.12
MDL Number MFCD00190102
InChI Key LAPGMTOHOQPDGI-UHFFFAOYSA-N
Synonym 2,5-difluoro-4-aminobenzonitrile, 4-cyano-2,5-difluoroaniline, 4-amino-2,5-difluoro-benzonitrile, 4-amino-2,5-difluorobenzenecarbonitrile, 4-cyano, pubchem3536, ksc507o5b, 4-cyano-2,5-difluoro-aniline, timtec-bb sbb028311, buttpark 100\07-75
PubChem CID 2735900
IUPAC Name 4-amino-2,5-difluorobenzonitrile
SMILES C1=C(C(=CC(=C1F)N)F)C#N

Specifications

Melting Point 97°C
Color Yellow
Quantity 5 g
UN Number 3439
Formula Weight 154.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2,5-difluorobenzonitrile