missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,5-difluorobenzonitrile, 96%

Catalog No. AAB2361106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2361106 5 g
AAB2361103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2361106 Supplier Thermo Scientific Chemicals Supplier No. B2361106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112279-61-5 | C7H4F2N2 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112279-61-5
Molecular Formula C7H4F2N2
Molecular Weight (g/mol) 154.12
MDL Number MFCD00190102
InChI Key LAPGMTOHOQPDGI-UHFFFAOYSA-N
Synonym 2,5-difluoro-4-aminobenzonitrile, 4-cyano-2,5-difluoroaniline, 4-amino-2,5-difluoro-benzonitrile, 4-amino-2,5-difluorobenzenecarbonitrile, 4-cyano, pubchem3536, ksc507o5b, 4-cyano-2,5-difluoro-aniline, timtec-bb sbb028311, buttpark 100\07-75
PubChem CID 2735900
IUPAC Name 4-amino-2,5-difluorobenzonitrile
SMILES C1=C(C(=CC(=C1F)N)F)C#N

Specifications

Melting Point 91°C to 94°C
Quantity 5 g
UN Number UN3439
Formula Weight 154.12
Percent Purity 96%
Chemical Name or Material 4-Amino-2,5-difluorobenzonitrile
Hazard Category H301-H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only