missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-butanol, 98%

Catalog No. p-7020647
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1268003 1 g
AAA1268006 5 g
AAA1268014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1268003 Supplier Thermo Scientific Chemicals Supplier No. A1268003

CAS: 13325-10-5 | C4H11NO | 89.138 g/mol

4-Amino-1-butanol is a useful intermediate in a variety of organic synthesis. It is mainly used in personal-care products, preparation of water soluble cationic flocculants and ion exchange resins as well as in the preparation of beta-lactam antibiotics. It is used for the production of efficient anionic emulsifiers, nonionic polyethylene emulsions, water treatment, metal treatment and absorption of carbon dioxide gas. It is used as a pigment dispersion aid and curing agent in selected textile -resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-1-butanol is a useful intermediate in a variety of organic synthesis. It is mainly used in personal-care products, preparation of water soluble cationic flocculants and ion exchange resins as well as in the preparation of beta-lactam antibiotics. It is used for the production of efficient anionic emulsifiers, nonionic polyethylene emulsions, water treatment, metal treatment and absorption of carbon dioxide gas. It is used as a pigment dispersion aid and curing agent in selected textile -resins.

Solubility
Miscible with water.

Notes
Air Sensitive and hygroscopic. Incompatible with strong oxidizing agents and acids.

Chemical Identifiers

CAS 13325-10-5
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00008230
InChI Key BLFRQYKZFKYQLO-UHFFFAOYSA-N
Synonym 4-amino-1-butanol, 4-aminobutanol, 1-butanol, 4-amino, 4-amino-butan-1-ol, 4-hdyroxybutylamine, 4-hydroxybutylamine, 4-hydroxy-n-butylamine, butanolamine, 4-amino butanol, 4-amino-butanol
PubChem CID 25868
IUPAC Name 4-aminobutan-1-ol
SMILES C(CCO)CN

Specifications

Melting Point 16°C to 18°C
Density 0.967
Boiling Point 206°C
Flash Point 107°C (224°F)
Odor Amine-like
Refractive Index 1.461
Linear Formula H2N(CH2)4OH
Quantity 1 g
UN Number UN2735
Beilstein 1731411
Sensitivity Air sensitive; Hygroscopic
Solubility Information Miscible with water.
Formula Weight 89.14
Percent Purity 98%
Chemical Name or Material 4-Amino-1-butanol
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 236-364-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only