missing translation for 'onlineSavingsMsg'
Learn More

4-Amidinobenzamide Hydrochloride 92.0+%, TCI America™

Catalog No. A106210G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A106210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A106210G Supplier TCI America Supplier No. A106210G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59855-11-7
Molecular Formula C8H10ClN3O
Molecular Weight (g/mol) 199.638
MDL Number MFCD00013011
InChI Key IYIAWOSYBLPUNL-UHFFFAOYSA-N
Synonym 4-amidinobenzamide hydrochloride, 4-carbamimidoylbenzamide hydrochloride, 4-amidino-benzoic acid amide hydrochloride, acmc-1awl9, 4-amidinobenzamide hcl, 4-carbamimidoyl-benzamide,hydrochloride, 4-amidinobenzoic acid amide hydrochloride, 4-amino imino methyl benzamide hydrochloride, benzamide, 4-aminoiminomethyl-, hydrochloride, benzamide,4-aminoiminomethyl-, hydrochloride 1:1
PubChem CID 2724293
IUPAC Name 4-carbamimidoylbenzamide;hydrochloride
SMILES C1=CC(=CC=C1C(=N)N)C(=O)N.Cl

Specifications

Color White
Quantity 10 g
Formula Weight 199.64
Percent Purity ≥92.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amidinobenzamide Hydrochloride