missing translation for 'onlineSavingsMsg'
Learn More

4-Allylanisole, 98%, Thermo Scientific™

Catalog No. AC102881000
Change view
Click to view available options
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC102881000 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC102881000 Supplier Thermo Scientific Chemicals Supplier No. 102881000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 140-67-0
CAS Min % 97.5
CAS Max % 100.0
Density 0.9600g/mL
Boiling Point 215°C to 216°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H12O
Refractive Index 1.5205 to 1.5225
Linear Formula H2C=CHCH2C6H4OCH3
Quantity 100g
MDL Number MFCD00008653
Beilstein 06, 571
Specific Gravity 0.96
Merck Index 15, 3760
Synonym estragole, 4-allylanisole, p-allylanisole, tarragon, methyl chavicol, 1-allyl-4-methoxybenzene, estragol, esdragol, esdragole, esdragon
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and chloroform
InChI Key ZFMSMUAANRJZFM-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)CC=C
IUPAC Name 1-methoxy-4-prop-2-enylbenzene
Molecular Weight (g/mol) 148.2
PubChem CID 8815
ChEBI CHEBI:4867
Formula Weight 148.2
Percent Purity 98%
Chemical Name or Material 4-Allylanisole, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing genetic defects.
Suspected of causing cancer.
Harmful to aqu
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER/doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/at
EINECSNumber 205-427-8
RTECSNumber BZ8225000
TSCA TSCA

RUO – Research Use Only