missing translation for 'onlineSavingsMsg'
Learn More

4-Allylanisole, 98%

Catalog No. AAB2331122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2331122 100 g
AAB2331114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2331122 Supplier Thermo Scientific Chemicals Supplier No. B2331122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 140-67-0 | C10H12O | 148.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 140-67-0
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00008653
InChI Key ZFMSMUAANRJZFM-UHFFFAOYSA-N
Synonym estragole, 4-allylanisole, p-allylanisole, tarragon, methyl chavicol, 1-allyl-4-methoxybenzene, estragol, esdragol, esdragole, esdragon
PubChem CID 8815
ChEBI CHEBI:4867
IUPAC Name 1-methoxy-4-prop-2-enylbenzene
SMILES COC1=CC=C(C=C1)CC=C

Specifications

Density 0.965
Boiling Point 215°C to 216°C
Flash Point 81°C (177°F)
Refractive Index 1.521
Quantity 100 g
Beilstein 1099454
Merck Index 14,3705
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material 4-Allylanisole
Hazard Category H227-H302-H315-H317-H319-H341-H351
Hazard Statement GHS H Statement
H301-H341-H351-H227-H315-H319-H335
Toxic if swallowed.
Suspected of causing genetic defects.
Suspected of causing cancer.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P261-P264b-P270-P272-P280-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P333+P313-P362-P370+P378q-P501c
EINECSNumber 205-427-8
RTECSNumber BZ8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only