missing translation for 'onlineSavingsMsg'
Learn More

4-Allyl-1,2-dimethoxybenzene 98.0+%, TCI America™

Catalog No. D1360500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1360500ML 500 mL
D136025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1360500ML Supplier TCI America Supplier No. D1360500ML
Only null left

Chemical Identifiers

CAS 93-15-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00008652
InChI Key ZYEMGPIYFIJGTP-UHFFFAOYSA-N
Synonym methyleugenol, methyl eugenol, 4-allyl-1,2-dimethoxybenzene, eugenol methyl ether, 4-allylveratrole, o-methyleugenol, eugenyl methyl ether, 3,4-dimethoxyallylbenzene, methyl eugenol ether, veratrole methyl ether
PubChem CID 7127
ChEBI CHEBI:4918
IUPAC Name 1,2-dimethoxy-4-prop-2-enylbenzene
SMILES COC1=C(C=C(C=C1)CC=C)OC

Specifications

Melting Point -4°C
Color Yellow
Boiling Point 120°C
Quantity 500 mL
Formula Weight 178.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Allyl-1,2-dimethoxybenzene
EINECSNumber (3)-0638
RTECSNumber CY2450000
TSCA Yes