missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acryloylmorpholine (stabilisée avec MEHQ) 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Boiling Point | 165°C |
| Quantity | 25 mL |
| Formula Weight | 141.17 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 4-Acryloylmorpholine (stabilized with MEHQ) |
Sécurité et manipulation
| EINECSNumber | (5)-0865 |
| TSCA | Yes |