missing translation for 'onlineSavingsMsg'
Learn More

4'-Acetylbenzo-15-crown 5-Ether 97.0+%, TCI America™

Catalog No. A16031G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16031G Supplier TCI America Supplier No. A16031G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41757-95-3
Molecular Formula C16H22O6
Molecular Weight (g/mol) 310.35
MDL Number MFCD00188564
InChI Key XNJYHEGDUAQGEE-UHFFFAOYSA-N
Synonym 2,3-(4-Acetylbenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene
PubChem CID 3038641
IUPAC Name 1-(2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecin-15-yl)ethan-1-one
SMILES CC(=O)C1=CC=C2OCCOCCOCCOCCOC2=C1

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 1 g
Formula Weight 310.35
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Acetylbenzo-15-crown 5-Ether
RTECSNumber KM5782540
TSCA No