missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxycinnamic acid, predominantly trans, 98+%

Catalog No. AAA1297906
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1297906 5 g
AAA1297922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1297906 Supplier Thermo Scientific Chemicals Supplier No. A1297906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15486-19-8 | C11H10O4 | 206.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15486-19-8
Molecular Formula C11H10O4
Molecular Weight (g/mol) 206.197
MDL Number MFCD00016847
InChI Key BYHBHNKBISXCEP-QPJJXVBHSA-N
Synonym 4-acetoxycinnamic acid, p-acetoxycinnamic acid, 3-4-acetoxyphenyl acrylic acid, 2-propenoic acid, 3-4-acetyloxy phenyl, trans-4-acetoxycinnamic acid, 3-4-acetyloxy phenyl prop-2-enoic acid, 4-acetoxycinnamic acid, predominantly trans, acetylated coumaric acid, 2e-3-4-acetyloxy phenyl prop-2-enoic acid, 3-4-acetoxyphenyl-2-propenoic acid
PubChem CID 5373941
ChEBI CHEBI:86580
IUPAC Name (E)-3-(4-acetyloxyphenyl)prop-2-enoic acid
SMILES CC(=O)OC1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 205°C to 208°C
Odor Odorless
Quantity 5 g
Beilstein 2645987
Formula Weight 206.2
Percent Purity ≥98%
Chemical Name or Material 4-Acetoxycinnamic acid, predominantly trans
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-512-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only