missing translation for 'onlineSavingsMsg'
Learn More

4'-Acetamidoacetophenone, 98%

Catalog No. AAL0343018
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0343018 50 g
AAL0343009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0343018 Supplier Thermo Scientific Chemicals Supplier No. L0343018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2719-21-3 | C10H11NO2 | 177.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2719-21-3
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.203
MDL Number MFCD00014965
InChI Key WECHHDJTILFYQT-UHFFFAOYSA-N
Synonym 4'-acetamidoacetophenone, n-4-acetylphenyl acetamide, 4-acetamidoacetophenone, n-p-acetylphenyl acetamide, acetamide, n-4-acetylphenyl, p-acetylaminoacetophenone, 4-acetylacetanilide, 4'-acetylacetanilide, p-acetamidoacetophenone, 4-acetamido-acetophenone
PubChem CID 75937
IUPAC Name N-(4-acetylphenyl)acetamide
SMILES CC(=O)C1=CC=C(C=C1)NC(=O)C

Specifications

Melting Point 166°C to 172°C
Quantity 50 g
Beilstein 639749
Formula Weight 177.2
Percent Purity 98%
Chemical Name or Material 4'-Acetamidoacetophenone
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 220-320-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only