missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamido-2,2,6,6-tetramethyl-1-oxopiperidinium Tetrafluoroborate 95.0+%, TCI America™

Catalog No. A20655G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A20655G Supplier TCI America Supplier No. A20655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 219543-09-6
Molecular Formula C11H21BF4N2O2
Molecular Weight (g/mol) 300.105
InChI Key HTMHEICBCHCWAU-UHFFFAOYSA-O
PubChem CID 2753331
IUPAC Name N-(2,2,6,6-tetramethyl-1-oxopiperidin-1-ium-4-yl)acetamide;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CC(=O)NC1CC([N+](=O)C(C1)(C)C)(C)C

Specifications

Melting Point 181°C
Color Yellow
Quantity 5 g
UN Number 1759
Formula Weight 300.10
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamido-2,2,6,6-tetramethyl-1-oxopiperidinium Tetrafluoroborate