missing translation for 'onlineSavingsMsg'
Learn More

4',7-Dihydroxyisoflavone, 98+%

Catalog No. AAJ63763ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63763ME 500 mg
AAJ63763MC 100 mg
AAJ6376303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ63763ME Supplier Thermo Scientific Chemicals Supplier No. J63763ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analog of genistein and an agonist at estrogen recpetors | CAS: 486-66-8 | C15H10O4 | 254.241 g/mol

Analog of genistein and an agonist at estrogen recpetors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 486-66-8
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.241
MDL Number MFCD00016954
InChI Key ZQSIJRDFPHDXIC-UHFFFAOYSA-N
Synonym daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone
PubChem CID 5281708
ChEBI CHEBI:28197
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
SMILES C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O

Specifications

Melting Point 328°C to 332°C (decomposition)
Color White to Yellow
Quantity 500 mg
Beilstein 231523
Merck Index 14,2801
Formula Weight 254.24
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 4',7-Dihydroxyisoflavone
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 207-635-4
RTECSNumber DJ3100040
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only