missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine 97.0+%, TCI America™

Catalog No. D4442200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4442200MG 200 mg
D44421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4442200MG Supplier TCI America Supplier No. D4442200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 333432-27-2
Molecular Formula C5HBr2N3S
Molecular Weight (g/mol) 294.952
MDL Number MFCD23703147
InChI Key LEHZIBSAFRVAJP-UHFFFAOYSA-N
PubChem CID 58129789
IUPAC Name 4,7-dibromo-[1,2,5]thiadiazolo[3,4-c]pyridine
SMILES C1=C(C2=NSN=C2C(=N1)Br)Br

Specifications

Melting Point 148°C
Color Yellow
Quantity 200 mg
Formula Weight 294.95
Percent Purity ≥97.0% (GC)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine
TSCA No
Recommended Storage Refrigerator