missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,7-Dibromo-1,10-phenanthroline Hydrate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 236°C |
| Color | Yellow |
| Quantity | 200 mg |
| Formula Weight | 338.00 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,7-Dibromo-1,10-phenanthroline Hydrate |
Safety and Handling
| TSCA | No |