missing translation for 'onlineSavingsMsg'
Learn More

4,7-Di(2-thienyl)-2,1,3-benzothiadiazole 98.0+%, TCI America™

Catalog No. D4487200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4487200MG 200 mg
D44871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4487200MG Supplier TCI America Supplier No. D4487200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 165190-76-1
Molecular Formula C14H8N2S3
Molecular Weight (g/mol) 300.412
MDL Number MFCD18252231
InChI Key XGERJWSXTKVPSV-UHFFFAOYSA-N
PubChem CID 396294
IUPAC Name 4,7-dithiophen-2-yl-2,1,3-benzothiadiazole
SMILES C1=CSC(=C1)C2=CC=C(C3=NSN=C23)C4=CC=CS4

Specifications

Melting Point 124°C
Quantity 200 mg
Formula Weight 300.41
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Di(2-thienyl)-2,1,3-benzothiadiazole