missing translation for 'onlineSavingsMsg'
Learn More

{4-[(6-Methylpyrazin-2-yl)oxy]phenyl}methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC62709DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC62709DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

906353-02-4
216.24
RNFIYSOVNHRFJW-UHFFFAOYSA-N
24229691
CC1=CN=CC(OC2=CC=C(CO)C=C2)=N1
C12H12N2O2
MFCD09817533
4-6-methylpyrazin-2-yl oxy phenyl methanol, 4-6-methylpyrazin-2-yl oxy benzyl alcohol, 4-6-methylpyrazin-2-yloxy phenyl methan-1-ol
[4-(6-methylpyrazin-2-yl)oxyphenyl]methanol

Specifications

{4-[(6-Methylpyrazin-2-yl)oxy]phenyl}methanol
96.5
97%
C12H12N2O2
MFCD09817533
RNFIYSOVNHRFJW-UHFFFAOYSA-N
[4-(6-methylpyrazin-2-yl)oxyphenyl]methanol
24229691
97%
906353-02-4
100.0
Amber Glass Bottle
1g
4-6-methylpyrazin-2-yl oxy phenyl methanol, 4-6-methylpyrazin-2-yl oxy benzyl alcohol, 4-6-methylpyrazin-2-yloxy phenyl methan-1-ol
CC1=CN=CC(OC2=CC=C(CO)C=C2)=N1
216.24
216.24
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents